Learn More
Raybiotech Inc Human IL-17 RC ELISA Kit
Supplier:  Raybiotech Inc ELHIL17RC1
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 0.05 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human IL-17RC. Other species not determined.
Catalog No. 50-194-9408