Learn More
TRIBIOSCIENCE INC Human IL-18 ELISA, 1 x 96-well plate
SDP
Catalog No. NC3454592
Encompass_Preferred
Tribioscience’s Human IL-18 ELISA is designed to quantitatively detect human IL-18 levels in in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 8-2000 pg/mL.
Catalog No. NC3454592 Supplier TRIBIOSCIENCE INC Supplier No. TBS3239
Tribioscience’s Human IL-18 ELISA is designed to quantitatively detect human IL-18 levels in in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 8-2000 pg/mL.