Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-18 ELISA Kit

Catalog No. 5018197
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-181-97 10 x 96 Tests
50-181-94 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-181-97 Supplier Invitrogen™ Supplier No. BMS2672TEN
  • $5,240.00
    $4,925.65 / Each of 1

    Save $314.35

    6% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-18
Biotin-Conjugate anti-human IL-18 monoclonal antibody
Streptavidin-HRP
Human IL-18 Standard lyophilized, 10ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

ELISA

The Human Interleukin-18 (Hu IL-18) ELISA quantitates Hu IL-18 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-18. Principle of the method The Human IL-18 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14116
Assay Range 78-5000 pg/mL
Assay Sensitivity 9.0 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320; IGIF; IL-18; IL-1g; IL1F4
Gene ID (Entrez) 3606
Gene Symbol IL18
Interassay CV 8.1%
Intraassay CV 6.5%
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Controls, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Controls, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2-8°C
Synonym Interleukin-18, Interferon-gamma-inducing factor
Target IL-18
Test Time 1 hr 20 min
Total Assay Time 3 hr 10 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.