Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems ACROBiosystems Human IL-18 Protein, Premium Grade, Recombinant, 50 μg

Catalog No. NC2893804
Encompass_Preferred

Recombinant human interleukin-18 (IL-18) premium-grade protein produced in E. coli and supplied lyophilized for preclinical research and cell culture applications. The protein stimulates IFN-γ secretion by KG-1 cells, offers high specific activity and low endotoxin, and is intended for discovery and process development.

  • Includes 50 μg of lyophilized recombinant human IL-18.
  • Expressed in E. coli cells.
  • Contains amino acids Tyr 37-Asp 193 (accession Q14116-1).
  • Specific activity greater than 3.00x10^6 IU/mg (WHO-calibrated).
  • Purity greater than 95%
  • Endotoxin less than 0.01 EU/μg by LAL or rFC.
  • Host cell protein <0.5 ng/μg and host cell DNA <0.02 ng/μg.
  • Sterility negative; mycoplasma negative.
  • Lyophilized from PBS (pH 7.4) with trehalose; store lyophilized at -20°C to -70°C for up to 24 months; after reconstitution store at -70°C for up to 3 months; avoid repeated freeze-thaw cycles.

Catalog No. NC2893804 Supplier ACROBiosystems Supplier No. IL8H511450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interleukin-18 (IL-18) premium-grade protein produced in E. coli and supplied lyophilized for preclinical research and cell culture applications. The protein stimulates IFN-γ secretion by KG-1 cells, offers high specific activity and low endotoxin, and is intended for discovery and process development.

  • Includes 50 μg of lyophilized recombinant human IL-18.
  • Expressed in E. coli cells.
  • Contains amino acids Tyr 37-Asp 193 (accession Q14116-1).
  • Specific activity greater than 3.00x10^6 IU/mg (WHO-calibrated).
  • Purity greater than 95%
  • Endotoxin less than 0.01 EU/μg by LAL or rFC.
  • Host cell protein <0.5 ng/μg and host cell DNA <0.02 ng/μg.
  • Sterility negative; mycoplasma negative.
  • Lyophilized from PBS (pH 7.4) with trehalose; store lyophilized at -20°C to -70°C for up to 24 months; after reconstitution store at -70°C for up to 3 months; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.