Learn More
ACROBiosystems Human IL-18 R beta / IL-1 R7, Fc Tag (ILB-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Arg 356 (Accession # O95256-1).
Catalog No. 502230661
Encompass_Preferred
IL-18 R beta (IL-1R7), also known as IL18RAP (Interleukin 18 Receptor Accessory Protein), is a member of the IL-1 family. Within the IL18 receptor complex, does not mediate IL18-binding, but involved in IL18-dependent signal transduction, leading to NF-kappa-B and JNK activation
This item is not returnable. View return policy
Catalog No. 50-223-0661 Supplier ACROBiosystems Supplier No. ILBH5256100UG
IL-18 R beta (IL-1R7), also known as IL18RAP (Interleukin 18 Receptor Accessory Protein), is a member of the IL-1 family. Within the IL18 receptor complex, does not mediate IL18-binding, but involved in IL18-dependent signal transduction, leading to NF-kappa-B and JNK activation