Learn More
ACROBiosystems Biotinylated Human IL-18 R1, His,Avitag (IL1-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Arg 329 (Accession # Q13478-1).
Supplier:  ACROBiosystems IL1H82E725UG
Encompass_Preferred
Interleukin-18 receptor 1 (IL18R1) is also known as CD218 antigen-like family member A (CD218a), IL1 receptor-related protein (IL-1Rrp or IL1R-rp), CDw218a, IL18Ra, IL1RRP, which belongs to the interleukin-1 receptor family. IL18R1 contains three Ig-like C2-type (immunoglobulin-like) domains and one TIR domain. IL18R1 is receptor for interleukin 18 (IL-18). IL18R1 binds to the agonist leads to the activation of NF-kappa-B, but does not bind IL1A/interleukin-1 alpha or IL1B/interleukin-1 beta.
Catalog No. 50-225-3345