Learn More
ACROBiosystems ACROBiosystems Human IL-18BP Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems ILPH52221MG
Encompass_Preferred
Recombinant Protein;1MG;Human IL-18BP, His Tag (ILP-H5222) is expressed from human 293 cells (HEK293). It contains AA Thr 31 - Gly 194 (Accession # NP_001138529).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.5 kDa. The protein migrates as 32-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Human IL-18BP, His Tag (Cat. No. ILP-H5222) inhibits the IL-18-induced IFN-gamma secretion by KG‑1 cells. The ED50 for this effect is 0.019-0.053 µg/mL in the presence of 30 ng/mL of Recombinant Human IL‑18 (Routinely tested).
Catalog No. 50-210-6681