Learn More
Sino Biological Human IL-18Rα Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785767
Encompass_Preferred
The human IL18R1 / CD218a ELISA Pair Set is for the quantitative determination of human IL18R1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for IL18R1 /CD218a coated on a 96-well plate. Standards and samples are added to the wells,and any IL18R1 / CD218a present binds to the immobilized antibody. The wellsare washed and a horseradish peroxidase conjugated mouse anti-IL18R1 / CD218amonoclonal antibody is then added, producing an antibody-antigen-antibody"sandwich". The wells are again washed and TMB substrate solution is loaded,which produces color in proportion to the amount of IL18R1 / CD218a present inthe sample. To end the enzyme reaction, the stop solution is added andabsorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85767 Supplier Sino Biological Supplier No. SEK111025
The human IL18R1 / CD218a ELISA Pair Set is for the quantitative determination of human IL18R1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for IL18R1 /CD218a coated on a 96-well plate. Standards and samples are added to the wells,and any IL18R1 / CD218a present binds to the immobilized antibody. The wellsare washed and a horseradish peroxidase conjugated mouse anti-IL18R1 / CD218amonoclonal antibody is then added, producing an antibody-antigen-antibody"sandwich". The wells are again washed and TMB substrate solution is loaded,which produces color in proportion to the amount of IL18R1 / CD218a present inthe sample. To end the enzyme reaction, the stop solution is added andabsorbances of the microwell are read at 450 nm.