Learn More
FUJIFILM BIOSCIENCES INC Human IL-19, Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 1008810UG
Encompass_Preferred
Interleukin-19 (IL-19) is a member of the interleukin 10 (IL-10) cytokine family and is produced by B cells and monocytes. IL-19 binds the interleukin 20 receptor complex (IL-20R) to activate STAT3 signaling. IL-19 induces interleukin 6 (IL-6) and tumor necrosis factor alpha (TNF-a) expression in monocytes, and promotes T helper 2 (Th2) cell-mediated immune responses. IL-19 production is up-regulated in resting monocytes following granulocyte-macrophage colony-stimulating factor (GM-CSF) or lipopolysaccharide (LPS) stimulation.
Catalog No. 50-990-781