Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-19 Recombinant Protein, PeproTech®

Catalog No. 20019100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20019100UG 100 μg
200-19-10UG 10 μg
200-19-1MG 1 mg
20019250UG 250 μg
200-19-2UG 2 μg
20019500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 20019100UG Supplier Gibco™ Supplier No. 20019100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-19 is a 35.8 kDa homodimer of two 154 amino acid chains. In solution, IL-19 exists predominantly as a non-disulfide-linked dimer. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a cytokine that belongs to the IL10 cytokine subfamily. This cytokine is found to be preferentially expressed in monocytes. It can bind the IL20 receptor complex and lead to the activation of the signal transducer and activator of transcription 3 (STAT3). A similar cytokine in mouse is reported to up-regulate the expression of IL6 and TNF-alpha and induce apoptosis, which suggests a role of this cytokine in inflammatory responses. Alternatively spliced transcript variants encoding the distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UHD0
For Use With (Application) Control, Western Blot Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 29949
Molecular Weight (g/mol) 35.8 kDa
Name Human IL-19
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias il 19; IL-10C; IL19; IL-19; ILN; Interleukin; interleukin 19; Interleukin19; interleukin-19; MDA1; MDA-1; melanoma differentiation associated protein-like protein; melanoma differentiation-associated protein-like protein; NG.1; ZMDA1
Common Name IL-19
Gene Symbol IL19
Biological Activity Determined by its ability to stimulate the proliferation of HEK-Blue IL-19 TM reporter cells.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MLRRCLISTD MHHIEESFQE IKRAIQAKDT FPNVTILSTL ETLQIIKPLD VCCVTKNLLA FYVDRVFKDH QEPNPKILRK ISSIANSFLY MQKTLRQCQE QRQCHCRQEA TNATRVIHDN YDQLEVHAAA IKSLGELDVF LAWINKNHEV MSSA
Content And Storage -20°C
Activity Determined by its ability to stimulate the proliferation of HEK-Blue IL-19TM reporter cells.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.