Learn More
Raybiotech Inc Human IL-36 Ra (IL-1 F5) ELISA Kit
Supplier:  Raybiotech Inc ELHIL1F51
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.07 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 RA (IL-1 F3), IL-18 (IL-1 F4), IL-36 alpha (IL-1 F6), IL-37 (IL-1 F7), IL-36 beta (IL-1 F8), IL-36 gamma (IL-1 F9), IL-38 (IL-1 F10), IL-33 (IL-1 F11)
Catalog No. 50-194-9415