Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-1ra/IL-1F3 Quantikine ELISA Kit

Catalog No. SRA00B
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SRA00B 6 Plates
DRA00B 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SRA00B Supplier R&D Systems™ Supplier No. SRA00B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human IL-1ra/IL-1F3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-1ra/IL-1F3 Quantikine ELISA Kit quantifies human IL-1ra/IL-1F3/IL1RN in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 18.3pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Human IL-1 ra
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3557
Quantity 6 Plates
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym DIRA, ICIL-1 RA, IL1F3, IL-1F3, IL-1 ra, IL1RA, IL-1 ra3, IL1RN, IL-1 RN, interleukin 1 receptor antagonist, IRAP, MVCD4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.