Learn More
ACROBiosystems ACROBiosystems Human IL-1RL1 / ST2 Protein, His Tag
Catalog No. 502106664
Encompass_Preferred
Recombinant Protein;200UG;Human IL-1RL1, His Tag (IL1-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Phe 328 (Accession # Q01638-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.9 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1RL1, His Tag (Cat. No. IL1-H5229) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-33, His,Avitag (Cat. No. IL3-H82H5) with a linear range of 0.02-0.313 μg/mL (QC tested).;Immobilized human IL33 at 5 μg/mL (100 μL/well) can bind Human IL-1RL1, His Tag (Cat. No. IL1-H5229) with a linear range of 0.125-0.5 μg/mL (Routinely tested).
Catalog No. 50-210-6664 Supplier ACROBiosystems Supplier No. IL1H5229200UG
Recombinant Protein;200UG;Human IL-1RL1, His Tag (IL1-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Phe 328 (Accession # Q01638-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.9 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1RL1, His Tag (Cat. No. IL1-H5229) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-33, His,Avitag (Cat. No. IL3-H82H5) with a linear range of 0.02-0.313 μg/mL (QC tested).;Immobilized human IL33 at 5 μg/mL (100 μL/well) can bind Human IL-1RL1, His Tag (Cat. No. IL1-H5229) with a linear range of 0.125-0.5 μg/mL (Routinely tested).