Learn More
ACROBiosystems Human IL-2 R beta&IL-2 R gamma Heterodimer Protein, Fc Tag&Fc Tag (MALS verified)
Catalog No. 502055278
Encompass_Preferred
Recombinant Protein;100UG;Human IL-2RB&IL-2RG Heterodimer Protein, Fc Tag&Fc Tag (MALS verified) (ILG-H5254) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (IL-2RB) & Leu 23 - Asn 254 (IL-2RG) (Accession # P14784-1 (IL-2RB) & P31785-1 (IL-2RG)).;Human IL-2RB&IL-2RG Heterodimer Protein, Fc Tag&Fc Tag is produced by co-expression of IL-2RB and IL-2RG, has a calculated MW of 51.1 kDa (IL-2RB) and 53.6 kDa (IL-2RG). Subunit IL-2RB is fused with a human IgG1 Fc tag at the C-terminus and subunit IL-2RG is fused with a human IgG1 Fc tag at the C-terminus. The reducing (R) protein migrates as 65-80 kDa and 90-100 kDa respectively due to glycosylation.;Immobilized Human IL-2, Tag Free (Cat. No. IL2-H4113) at 5 μg/mL (100 μL/well) can bind Human IL-2RB&IL-2RG Heterodimer Protein, Fc Tag&Fc Tag (MALS verified) (Cat. No. ILG-H5254) with a linear range of 5-78 ng/mL (QC tested).;Immobilized Human IL-15, Tag Free (Cat. No. IL5-H4117) at 5 μg/mL (100 μL/well) can bind Hum
Catalog No. 50-205-5278 Supplier ACROBiosystems Supplier No. ILGH5254100UG
Recombinant Protein;100UG;Human IL-2RB&IL-2RG Heterodimer Protein, Fc Tag&Fc Tag (MALS verified) (ILG-H5254) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (IL-2RB) & Leu 23 - Asn 254 (IL-2RG) (Accession # P14784-1 (IL-2RB) & P31785-1 (IL-2RG)).;Human IL-2RB&IL-2RG Heterodimer Protein, Fc Tag&Fc Tag is produced by co-expression of IL-2RB and IL-2RG, has a calculated MW of 51.1 kDa (IL-2RB) and 53.6 kDa (IL-2RG). Subunit IL-2RB is fused with a human IgG1 Fc tag at the C-terminus and subunit IL-2RG is fused with a human IgG1 Fc tag at the C-terminus. The reducing (R) protein migrates as 65-80 kDa and 90-100 kDa respectively due to glycosylation.;Immobilized Human IL-2, Tag Free (Cat. No. IL2-H4113) at 5 μg/mL (100 μL/well) can bind Human IL-2RB&IL-2RG Heterodimer Protein, Fc Tag&Fc Tag (MALS verified) (Cat. No. ILG-H5254) with a linear range of 5-78 ng/mL (QC tested).;Immobilized Human IL-15, Tag Free (Cat. No. IL5-H4117) at 5 μg/mL (100 μL/well) can bind Hum