Learn More
ACROBiosystems ACROBiosystems Human IL-2 R gamma / CD132 Protein, His Tag (MALS verified)
Catalog No. 502107172
Encompass_Preferred
Recombinant Protein;100UG;Human IL-2 R gamma, His Tag (ILA-H52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asn 254 (Accession # P31785-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.3 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7172 Supplier ACROBiosystems Supplier No. ILAH52H5100UG
Recombinant Protein;100UG;Human IL-2 R gamma, His Tag (ILA-H52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asn 254 (Accession # P31785-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.3 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.