Learn More
ACROBiosystems Recombinant Protein;Human IL-2 R gamma / CD132 Protein, Fc Tag;HEK293;100UG;ILG,IL2RG,CD132,CIDX,IL-2RG,IMD4,P64,SCIDX,SCIDX1,gammaC,IL-2,IL2,IL-4
Supplier:  ACROBiosystems ILGH5256100UG
Encompass_Preferred
Recombinant Protein;Human IL-2 R gamma / CD132 Protein, Fc Tag;100UG;Human IL-2 R gamma, Fc Tag (ILG-H5256) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asn 254 (Accession # AAH14972).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53.5 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. NC1464354