Learn More
Raybiotech Inc Human IL-20 ELISA Kit
Supplier:  Raybiotech Inc ELHIL201
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 1 pg/ml . Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human IL-20. Other species not determined yet.
Catalog No. 50-194-9427