Learn More
FUJIFILM BIOSCIENCES INC Human IL-20, Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 1008910UG
Encompass_Preferred
Interleukin 20 (IL-20) is structurally related to interleukin 10 (IL-10) and is produced by keratinocytes and monocytes. IL-20 acts through the STAT3 signaling pathway to regulate the proliferation of keratinocytes during epidermal inflammation.
Catalog No. 50-900-478