Learn More
ACROBiosystems Human IL-20 R alpha Protein, His Tag (ILA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Val 30 - Lys 250 (Accession # Q9UHF4-1).
Catalog No. 502253178
Encompass_Preferred
IL-20 R alpha (Interleukin-20 receptor subunit alpha) forms a functional heterodimer with the IL-20 beta subunit, and transduces ligand-binding at least in part through STAT3 activation. The IL-20 subfamily of cytokines and receptors are members of the larger IL-10 family, which are grouped together based on their utilization of common receptor subunits, similarities in their target-cell profiles, and biological functions. This subfamily consists of the cytokines, IL-19, IL-20, IL-22, IL-24, and IL-26, as well as the receptors, IL-20RA, IL-20RB, IL-10RB, and IL-22RA1.
This item is not returnable. View return policy
Catalog No. 50-225-3178 Supplier ACROBiosystems Supplier No. ILAH52H3100UG
IL-20 R alpha (Interleukin-20 receptor subunit alpha) forms a functional heterodimer with the IL-20 beta subunit, and transduces ligand-binding at least in part through STAT3 activation. The IL-20 subfamily of cytokines and receptors are members of the larger IL-10 family, which are grouped together based on their utilization of common receptor subunits, similarities in their target-cell profiles, and biological functions. This subfamily consists of the cytokines, IL-19, IL-20, IL-22, IL-24, and IL-26, as well as the receptors, IL-20RA, IL-20RB, IL-10RB, and IL-22RA1.