Learn More
Sino Biological Human IL-20R alpha/IL-20RA Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786401
Encompass_Preferred
The human IL20RA / IL20R1 ELISA Pair Set is for the quantitative determination of human IL20RA / IL20R1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for IL20RA /IL20R1 coated on a 96-well plate. Standards and samples are added to the wells,and any IL20RA / IL20R1 present binds to the immobilized antibody. The wellsare washed and a horseradish peroxidase conjugated rabbit anti-IL20RApolyclonal antibody is then added, producing an antibody-antigen-antibody"sandwich". The wells are again washed and TMB substrate solution is loaded,which produces color in proportion to the amount of IL20RA / IL20R1 present inthe sample. To end the enzyme reaction, the stop solution is added andabsorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86401 Supplier Sino Biological Supplier No. SEK1039715
The human IL20RA / IL20R1 ELISA Pair Set is for the quantitative determination of human IL20RA / IL20R1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for IL20RA /IL20R1 coated on a 96-well plate. Standards and samples are added to the wells,and any IL20RA / IL20R1 present binds to the immobilized antibody. The wellsare washed and a horseradish peroxidase conjugated rabbit anti-IL20RApolyclonal antibody is then added, producing an antibody-antigen-antibody"sandwich". The wells are again washed and TMB substrate solution is loaded,which produces color in proportion to the amount of IL20RA / IL20R1 present inthe sample. To end the enzyme reaction, the stop solution is added andabsorbances of the microwell are read at 450 nm.