Learn More
ACROBiosystems Human IL-21, premium grade (IL1-H5213) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Ser 162 (Accession # Q9HBE4-1).
Supplier:  ACROBiosystems IL1H521350UG
Encompass_Preferred
Interleukin-21 (IL-21) is a secreted protein which belongs to the IL-15 / IL-21 family. Interleukin-21 / IL-21 belongs to a family of cytokines that bind to a composite receptor consisting of a private receptor (IL21R) and the common cytokine receptor gamma chain (gamma(C)). Interleukin-21 / IL-21 impacts a number of cell types, including CD8+ memory T cells, NK cells and subsets of CD4 memory T cells. The IL-21R is widely distributed on lympho-haematopoietic cells. IL-21 is a pleiotropic cytokine produced by CD4+ T cells in response to antigenic stimulation. Its action generally enhances antigen-specific responses of immune cells. IL-21 promotes the anti-tumor activity of CD8+ T-cells and NK cells. IL-21 exerts its effect through binding to a specific type I cytokine receptor, IL-21R, which also contains the γ chain (γc) found in other cytokine receptors including IL-2, IL-4, IL-7, IL-9 and IL-15. The IL-21/IL-21R interaction triggers a cascade of events which includes activation of
Catalog No. 50-225-3084