Learn More
Raybiotech Inc Human IL-21 ELISA Kit
Supplier:  Raybiotech Inc ELHIL211
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 110 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human Adiponectin, Adipsin, AFP, AgRP, Angiogenin, Angiopoietin 1, Angiostatin, ANGPTL4, B2M, BCAM, CA125, CA15-3, Catheprin S, CD40, CEA, Cripto-1, CRP, DAN, DKK-1, E-Cadherin, EpCAM, ErbB2, Fas Ligand, Fcg RIIB/C, Follistatin, FSH, Galectin-7, GRO alpha, beta HCG, ICAM-2, IGF-1 sR, IL-1 sRII, IL-3, IL-13 R1, IL-13R2, IL-17B, IL-18 Rb, IL-2 Ra, IL-2 Rb, IL-23, LAP, Leptin, MMP-1, MMP-2, MMP-3, MMP-8, MMP-9, MMP-10, MMP-13, NCAM-1, Nidogen-1, NrCAM, NSE, OSM, PAI-1, PDGF-AB, Procalcitonin, Prolactin, PSA, Resistin, SDF-1 beta, sgp130, ShhN, Siglec-5, Siglec-9, ST2, TACE, TGF beta 2, Thyroglobulin, Tie-2, TIMP-4, TPO, TRAIL R4, TREM-1, TSH, VEGF-C, VEGFR1.
Catalog No. 50-194-9429