Learn More
Raybiotech Inc Human IL-21 R ELISA Kit
Supplier:  Raybiotech Inc ELHIL21R1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.61 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human IL-21 R.
Catalog No. 50-194-9430