Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-22 High Sensitivity Magnetic Luminex™ Performance Assay

Catalog No. lbhs5782
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LBHS5782 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. LBHS5782 Supplier R&D Systems™ Supplier No. LBHS5782
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Magnetic bead-based multiplex assay for the Luminex™ MAGPIX™ platform

  • Magnetic format allows for easier wash steps
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
  • Requires a small sample volume (<50 μL)
  • Run your assay in just 3-5 hours
  • Mass-calibrated standards for consistent results with every new lot of material
  • Enough reagents for one 96-well plate

Specifications

Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target IL-22
Bead Type Magnetic
Assay Sensitivity 0.529pg/mL
Assay Range 1.64 - 6700pg/mL (Serum, Heparin Plasma, EDTA Plasma), 1.64 - 1675pg/mL (Cell Culture Supernates)
Gene ID (Entrez) 50616
Gene Symbol IL22
Sample Type Serum, Plasma (EDTA, Heparin), Cell Culture Supernatant
Sample Volume Cell Culture Supernates (100μL),Serum (50μL),EDTA Plasma (50μL),Heparin Plasma (50μL)
Synonym Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Kit Contents
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Content And Storage Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.