Learn More
FUJIFILM BIOSCIENCES INC Human IL-22, Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 10091100UG
Encompass_Preferred
Interleukin 22 (IL-22), also called IL-TIF, is an IL-10 family member that is produced by activated dendritic cells and T lymphocytes.  IL-22 signals via a heteroduplex receptor consisting of IL-22R and IL-10RB chains.  IL-22 is a potent mediator of cellular inflammatory responses.  
Catalog No. 50-990-790