Learn More
ACROBiosystems Human IL-22 R alpha 1, Fc Tag (IL1-H5258) is expressed from human 293 cells (HEK293). It contains AA His 16 - Thr 228 (Accession # Q8N6P7-1).
Catalog No. 502253388
Encompass_Preferred
Interleukin-22 receptor subunit alpha-1(IL22RA1) is a component of the receptor for IL20, IL22 and IL24.Component of IL22 receptor formed by IL22RA1 and IL10RB enabling IL22 signaling via JAK/STAT pathways. IL22 also induces activation of MAPK1/MAPK3 and Akt kinases pathways. Component of one of the receptor for IL20 and IL24 formed by IL22RA1 and IL20RB also signaling through STATs activation. Mediates IL24 antiangiogenic activity as well as IL24 inhibitory effect on endothelial cell tube formation and differentiation.
This item is not returnable. View return policy
Catalog No. 50-225-3388 Supplier ACROBiosystems Supplier No. IL1H5258100UG
Interleukin-22 receptor subunit alpha-1(IL22RA1) is a component of the receptor for IL20, IL22 and IL24.Component of IL22 receptor formed by IL22RA1 and IL10RB enabling IL22 signaling via JAK/STAT pathways. IL22 also induces activation of MAPK1/MAPK3 and Akt kinases pathways. Component of one of the receptor for IL20 and IL24 formed by IL22RA1 and IL20RB also signaling through STATs activation. Mediates IL24 antiangiogenic activity as well as IL24 inhibitory effect on endothelial cell tube formation and differentiation.