Learn More
Raybiotech Inc Human IL-22 R alpha 1 ELISA Kit
Supplier:  Raybiotech Inc ELHIL22RA11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 10.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human IL-22 R alpha 1.
Catalog No. 50-194-9433