Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-22 R alpha 1 ELISA Kit

Catalog No. EH266RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH266RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH266RBX10 Supplier Invitrogen™ Supplier No. EH266RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human IL-22 R alpha 1 quantitates human IL-22 R alpha 1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-22 R alpha 1.

Component of the receptor for IL20, IL22 and IL24. Component of IL22 receptor formed by IL22RA1 and IL10RB enabling IL22 signaling via JAK/STAT pathways. IL22 also induces activation of MAPK1/MAPK3 and Akt kinases pathways. Component of one of the receptor for IL20 and IL24 formed by IL22RA1 and IL20RB also signaling through STATs activation. Mediates IL24 antiangiogenic activity as well as IL24 inhibitory effect on endothelial cell tube formation and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 10.5-2500 pg/mL
Assay Sensitivity 10.5 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 9130219A07Rik; CRF2-9; Cytokine receptor class-II member 9; cytokine receptor family 2 member 9; ENSMUSG00000073746; IL-22 receptor subunit alpha-1; Il22r; IL-22R; IL22R1; Il22ra1; IL-22RA1; IL-22R-alpha-1; interleukin 22 receptor subunit alpha 1; interleukin 22 receptor, alpha 1; interleukin-22 receptor alpha chain; interleukin-22 receptor subunit alpha-1; RGD1559655; zcytoR119130219A07Rik; CRF2-9; Cytokine receptor class-II member 9; cytokine receptor family 2 member 9; ENSMUSG00000073746; IL-22 receptor subunit alpha-1; Il22r; IL-22R; IL22R1; Il22ra1; IL-22RA1; IL-22R-alpha-1; interleukin 22 receptor subunit alpha 1; interleukin 22 receptor, alpha 1; interleukin-22 receptor alpha chain; interleukin-22 receptor subunit alpha-1; RGD1559655; zcytoR11;
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IL22RA1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.