Learn More
ACROBiosystems ACROBiosystems Human IL-23 alpha & Rat IL-12 beta Heterodimer Protein, His Tag&Tag Free (MALS verified)
Catalog No. 502107388
Encompass_Preferred
Recombinant Protein;50UG;Human IL-23A & Rat IL-12B Heterodimer Protein, His Tag&Tag Free (ILB-HR52W3) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Pro 189 (IL23A) & Met 23 - Ser 335 (IL12B) (Accession # Q9NPF7-1 (IL23A) & Q9R278-1 (IL12B)).;Human IL-23A & Rat IL-12B Heterodimer Protein, His Tag&Tag Free, produced by co-expression of IL-23A and IL-12B, has a calculated MW of 20.6 kDa (IL-23A) and 35.9 kDa (IL-12B). Subunit IL-23A is fused with a polyhistidine tag at the N-terminus and subunit IL-12B contains no tag. The reducing (R) protein migrates as 23 kDa (IL-23A) and 45 kDa and 47 kDa (IL-12B) respectively due to glycosylation.;Immobilized Human IL-23 R, Fc Tag (Cat. No. ILR-H5254) at 5 μg/mL (100 μL/well) can bind Human IL-23A & Rat IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-HR52W3) with a linear range of 0.039-0.156 μg/mL (QC tested).;Immobilized Human IL-23A & Rat IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-HR52W3) at 5 μg/mL
Catalog No. 50-210-7388 Supplier ACROBiosystems Supplier No. ILBHR52W350UG
Recombinant Protein;50UG;Human IL-23A & Rat IL-12B Heterodimer Protein, His Tag&Tag Free (ILB-HR52W3) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Pro 189 (IL23A) & Met 23 - Ser 335 (IL12B) (Accession # Q9NPF7-1 (IL23A) & Q9R278-1 (IL12B)).;Human IL-23A & Rat IL-12B Heterodimer Protein, His Tag&Tag Free, produced by co-expression of IL-23A and IL-12B, has a calculated MW of 20.6 kDa (IL-23A) and 35.9 kDa (IL-12B). Subunit IL-23A is fused with a polyhistidine tag at the N-terminus and subunit IL-12B contains no tag. The reducing (R) protein migrates as 23 kDa (IL-23A) and 45 kDa and 47 kDa (IL-12B) respectively due to glycosylation.;Immobilized Human IL-23 R, Fc Tag (Cat. No. ILR-H5254) at 5 μg/mL (100 μL/well) can bind Human IL-23A & Rat IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-HR52W3) with a linear range of 0.039-0.156 μg/mL (QC tested).;Immobilized Human IL-23A & Rat IL-12B Heterodimer Protein, His Tag&Tag Free (Cat. No. ILB-HR52W3) at 5 μg/mL