Learn More
ACROBiosystems Human IL-23 / IL-23R Inhibitor Screening Kit (TR-FRET), 100 tests
Catalog No. NC3892239
Encompass_Preferred
The Human IL-23 / IL-23R Inhibitor Screening Kit (TR-FRET) is a homogeneous, no-wash competition assay for rapid screening of compounds or biologics that disrupt human IL-23 binding to IL-23R. It provides quantitative TR-FRET readouts in a 100-test format suitable for medium-throughput inhibitor screening and binding studies.
  • Competition TR-FRET assay in a homogeneous, no-wash format.
  • Includes europium-chelate labeled IL-23 heterodimer, FA-labeled IL-23R, neutralizing antibody, and buffers.
  • 100-test kit format with ~0.5-1 hour assay time.
  • Sample volume: 10 μL per reaction.
  • Store unopened kit at 2-8 °C; follow component instructions after opening.
  • Intended for research use only (RUO).
Catalog No. NC3892239 Supplier ACROBiosystems Supplier No. FRTP010100TESTS
The Human IL-23 / IL-23R Inhibitor Screening Kit (TR-FRET) is a homogeneous, no-wash competition assay for rapid screening of compounds or biologics that disrupt human IL-23 binding to IL-23R. It provides quantitative TR-FRET readouts in a 100-test format suitable for medium-throughput inhibitor screening and binding studies.
  • Competition TR-FRET assay in a homogeneous, no-wash format.
  • Includes europium-chelate labeled IL-23 heterodimer, FA-labeled IL-23R, neutralizing antibody, and buffers.
  • 100-test kit format with ~0.5-1 hour assay time.
  • Sample volume: 10 μL per reaction.
  • Store unopened kit at 2-8 °C; follow component instructions after opening.
  • Intended for research use only (RUO).