Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-23 ELISA Kit

Catalog No. 501125254
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-5254 96 Tests
50-179-02 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-5254 Supplier Invitrogen™ Supplier No. BMS20233
Add to Cart
Add to Cart

Includes

Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-23
Biotin-Conjugate anti-human IL-23 monoclonal antibody
Avidin-HRP
Human IL-23 Standard, lyophilized, 4000pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

ELISA

The Human Interleukin-23 (Hu IL-23) ELISA quantitates Hu IL-23 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-23. Principle of the method The Human IL-23 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 that is disulfide-linked with to two subunits, a p19 subunit unique to IL-23, and a p40 subunit that is shared with IL-12. IL-23 interacts with IL-12Rbeta1 and a beta2-like receptor subunit with STAT4 binding domain, IL-23R. IL-23 is secreted by activated mouse and human dendritic cells and is involved in the inflammation process and associated with autoimmune diseases. Mouse IL-23 was found not to induce significant amounts of IFN-gamma, induces strong proliferation of memory T cells (but not naive T cells), and activate mouse memory T cells to produce the proinflammatory cytokine IL-17. On the other hand, human IL-23 induces proliferation of memory T cells, induces moderate levels of IFN-gamma, and is produced by naive and memory T cells. IL-23 plays an important role in stimulating memory T-cells, and is required for the induction, expansion, maintenance, and downstream effector functions of Th17 cells, which play a vital role in upregulating neutrophil chemokines and various pro-inflammatory cytokines. IL-23 interacts with the receptors IL-12R beta1 and IL-23R to activate the Jak-Stat signaling cascade. IL-23 has been shown to possess potent anti-tumor and anti-metastatic activity in mouse models of cancer, suggesting a potential role for IL-23 in therapeutic treatment of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NPF7
Assay Range 15.6-2000 pg/mL
Assay Sensitivity 4.0 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias IL23; IL-23; IL-23 subunit alpha; IL23A; IL-23A; IL-23-A; IL23P19; IL-23p19; ILN; Interleukin; interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; P19; SGRF; UNQ2498/PRO5798IL23; IL-23; IL-23 subunit alpha; IL23A; IL-23A; IL-23-A; IL23P19; IL-23p19; ILN; Interleukin; interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; P19; SGRF; UNQ2498/PRO5798; IL-23; IL-23A; IL23P19; P19; SGRF
Gene ID (Entrez) 51561
Gene Symbol IL23A
Interassay CV 6.3%
Intraassay CV 5.9%
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases IL-23 subunit alpha, IL-23-A, Interleukin-23 subunit p19, IL-23p19
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2-8°C
Synonym JKA3 induced upon T cell activation, SGRF
Target IL-23
Test Time 1 hr 20 min
Total Assay Time 3 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.