Learn More
Bon Opus Biosciences Recombinant Human IL-23R Domain1 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP108350UG
Encompass_Preferred
Recombinant Human IL-23R Domain1 (C-hFc) is produced by Human Cells. The target gene encoding Gly24-Tyr123 is expressed with a C-hFc tag. Accession# Q5VWK5-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1083
Catalog No. 50-217-2681