Learn More
Raybiotech Inc Human IL-26 (AK155) ELISA Kit
Supplier:  Raybiotech Inc ELHIL261
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 3.3 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human IL-26. Other species not yet determined.
Catalog No. 50-194-9437