Learn More
Invitrogen™ Human IL-26 ELISA Kit
Description
Human IL-26 quantitates human IL-26 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-26.
Specifications
Specifications
| Accession Number | Q9NPH9 |
| Assay Range | 3.3-800 pg/mL |
| Assay Sensitivity | 3.3 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | AK155; il 26; IL26; IL-26; ILN; Interleukin; interleukin 26; interleukin-26; Protein AK155AK155; il 26; IL26; IL-26; ILN; Interleukin; interleukin 26; interleukin-26; Protein AK155; |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.