Learn More
Raybiotech Inc Human IL-27 ELISA Kit
Supplier:  Raybiotech Inc ELHIL271
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 81 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human aFGF, Albumin, AMICA, Angiopoietin-4, BAFF, CD163, Clusterin, CRTAM, CXCL14, Cystatin C, DLL1, Fetuin A, FOLR1, GASP-1, GASP-2, G-CS R, HAI-2, IL-17B R, LAG-3, LDL R, Pepsinogen I, RBP4, SOST, Syndecan-1, TACI, TFPI, Thrombospondin-1, TRANCE, Troponin I, uPA, WISP-1.
Catalog No. 50-194-9438