Learn More
Abcam Human IL-27 ELISA Kit (IL-27 p28), 1 x 96 Tests
Catalog No. 502656456
Encompass_Preferred
Human IL-27 ELISA Kit (IL-27 p28) is a single-wash 90-min Simplestep used to quantify Human IL-27 (IL-27 p28) with a sensitivity of 61.02 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6456 Supplier Abcam Supplier No. AB2678121X96TESTS
Human IL-27 ELISA Kit (IL-27 p28) is a single-wash 90-min Simplestep used to quantify Human IL-27 (IL-27 p28) with a sensitivity of 61.02 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement