Learn More
Raybiotech Inc Human IL-27 R alpha ELISA Kit
Supplier:  Raybiotech Inc ELHIL27RA1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.85 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human IL-27 R alpha.
Catalog No. 50-194-9439