Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-27 R alpha ELISA Kit

Catalog No. EH272RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH272RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH272RBX10 Supplier Invitrogen™ Supplier No. EH272RBX10
Only null left
Add to Cart
Add to Cart
Request a Quote

ELISA

In mice, CD4+ helper T-cells differentiate into type 1 (Th1) cells and type 2 (Th2) cells, which are critical for cell-mediated immunity and most antibody responses, respectively. The differentiation of Th1 cells is predominantly under the influence of IL12, while IL4 influences the differentiation of Th2 cells. Mice deficient in these cytokines, their receptors, or associated transcription factors have impaired, but not absent, Th1 or Th2 immune responses. The gene discussed here encodes a protein that is similar to the mouse T-cell cytokine receptor Tccr at the amino acid level and is predicted to be a glycosylated transmembrane protein. Diseases associated with IL27RA include Hyper Ige Recurrent Infection Syndrome 4 and Crisponi/Cold-Induced Sweating Syndrome 2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6UWB1
Assay Range 0.85 to 200 ng/mL
Assay Sensitivity 0.85 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 9466
Gene Symbol Il27ra
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target TCCR
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.