Learn More
ACROBiosystems Human ANGPTL3 ELISA Kit, Sandwich-ELISA, 96 Tests
Catalog No. 502719764
Encompass_Preferred
The Human ANGPTL3 ELISA Kit, PRO is a ready-to-use sandwich enzyme immunoassay designed for the quantitative detection of natural and recombinant human ANGPTL3 in biological samples, including serum, plasma, and cell culture supernatants. This kit utilizes high-quality antibody pairs and protein standards to ensure consistent, reliable results with high sensitivity.
  • Calibrated to NIBSC/WHO standards for comparable results.
  • Validated for detection range, sensitivity, precision, recovery, and linearity according to ICH M10 guidelines.
  • Ready-to-use reagents and simplified protocols designed to save assay time.
  • Dynamic detection range of 0.156 ng/mL to 10 ng/mL with a sensitivity of 0.156 ng/mL.
  • Compatible with standard microplate readers measuring absorbance at 450 nm.
Catalog No. 50-271-9764 Supplier ACROBiosystems Supplier No. CEAB08996TESTS
The Human ANGPTL3 ELISA Kit, PRO is a ready-to-use sandwich enzyme immunoassay designed for the quantitative detection of natural and recombinant human ANGPTL3 in biological samples, including serum, plasma, and cell culture supernatants. This kit utilizes high-quality antibody pairs and protein standards to ensure consistent, reliable results with high sensitivity.
  • Calibrated to NIBSC/WHO standards for comparable results.
  • Validated for detection range, sensitivity, precision, recovery, and linearity according to ICH M10 guidelines.
  • Ready-to-use reagents and simplified protocols designed to save assay time.
  • Dynamic detection range of 0.156 ng/mL to 10 ng/mL with a sensitivity of 0.156 ng/mL.
  • Compatible with standard microplate readers measuring absorbance at 450 nm.