Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human R-Spondin 2 ELISA Kit

Catalog No. EH396RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH396RB 96 Tests
EH396RBX5 5 x 96 Tests
EH396RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH396RB Supplier Invitrogen™ Supplier No. EH396RB
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human R-Spondin 2 quantitates human R-Spondin 2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human R-Spondin 2.

The R-Spondin (Rspo) proteins belong to the Rspo family of Wnt modulators. Currently, the family consists of four structurally-related, secreted ligands (Rspo 1-4), all containing furin-like and thrombospondin structural domains. The Rspo proteins can interact with the Frizzled/LRP6 receptor complex in a manner that causes the stabilization, and resulting accumulation, of the intracellular signaling protein, beta-catenin. This activity effectively activates and increases the subsequent nuclear signaling of beta-catenin. R-Spondin can also bind to the previously discovered G-protein coupled receptors, LGR-4 and LGR-5. Rspo/beta-catenin signaling can act as an inducer of the transformed phenotype, and can also regulate the proliferation and differentiation of certain stem cell populations.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6UXX9
Assay Range 0.55-35 ng/mL
Assay Sensitivity 0.55 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 2610028F08Rik; AA673245; CRISTIN2; cristin-2; cysteine-rich and single thrombospondin domain-containing protein 2; D430027K22; ftls; hRspo2; mCristin-2; RGD1562331; Roof plate-specific spondin-2; Rspo2; R-spondin 2; R-spondin 2 homolog; R-spondin 2 homolog (Xenopus laevis); R-spondin-2; UNQ9384/PRO342092610028F08Rik; AA673245; CRISTIN2; cristin-2; cysteine-rich and single thrombospondin domain-containing protein 2; D430027K22; ftls; hRspo2; mCristin-2; RGD1562331; Roof plate-specific spondin-2; Rspo2; R-spondin 2; R-spondin 2 homolog; R-spondin 2 homolog (Xenopus laevis); R-spondin-2; UNQ9384/PRO34209;
Gene ID (Entrez) 340419
Gene Symbol RSPO2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target RSPO2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.