Learn More
ACROBiosystems Human IL-3, His Tag (IL3-H52H9) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Phe 152 (Accession # P08700-1).
Supplier:  ACROBiosystems IL3H52H9100UG
Encompass_Preferred
Interleukin 3 is also known as IL-3, is a protein that in humans is encoded by the IL3 gene. Interleukin-3 (IL-3) is an interleukin, a type of biological signal (cytokine) that can improve the body's natural response to disease as part of the immune system. It acts by binding to the interleukin-3 receptor. The IL-3 receptor is a heterodimer of the IL-3 specific α-chain and the common β-chain, βc, which is also used by GM-CSF and IL-5. [3] Binding of IL-3 can also involve substitution of the βc by a βIL-3-chain that appears to be specific for IL-3 . Binding of IL-3 to its cognate receptor(s) induces activation of Jak2, phosphorylation of multiple Stats, and the PI3K/Akt pathway. IL-3 may play an important role in the development of airway inflammation associated with asthma. The human IL-3 gene encodes a protein 152 amino acids long, and the naturally occurring IL-3 is glycosylated.
Catalog No. 50-223-0749