Learn More
ACROBiosystems Human IL-3 R alpha 25tests
Catalog No. 502534595
Encompass_Preferred
APC-Labeled Human IL-3 R alpha Protein His Tag (ILA-HA2H8) is produced via conjugation of APC to Human IL-3 R alpha Protein His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human IL-3 R alpha Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Thr 19-Arg 305 (Accession # P26951-1).
Catalog No. 50-253-4595 Supplier ACROBiosystems Supplier No. ILAHA2H825TESTS
APC-Labeled Human IL-3 R alpha Protein His Tag (ILA-HA2H8) is produced via conjugation of APC to Human IL-3 R alpha Protein His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human IL-3 R alpha Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Thr 19-Arg 305 (Accession # P26951-1).