Learn More
ACROBiosystems PE-Labeled Human IL-3 R alpha, His Tag (ILA-HP2H8) is produced via site-specific conjugation of PE to Human IL-3 R alpha, His Tag under optimal conditions with a proprietary technology. Human IL-3 R alpha, His Tag is expressed from 293 cell
Catalog No. 502231511
Encompass_Preferred
Interleukin 3 receptor alpha (low affinity) (IL3RA), also known as CD123 (Cluster of Differentiation 123) is a 70-kD glycoprotein member of the hematopoietin receptor superfamily. This protein associates with a beta subunit common to the receptors for IL-5 and granulocyte-macrophage colony-stimulating factor (GM-CSF) to form a high-affinity receptor for IL-3. The interleukin-3 receptor α chain (CD123) has been identified as a potential immunotherapeutic target because it is overexpressed in AML compared with normal hematopoietic stem cells.
This item is not returnable. View return policy
Catalog No. 50-223-1511 Supplier ACROBiosystems Supplier No. ILAHP2H825TESTS
Interleukin 3 receptor alpha (low affinity) (IL3RA), also known as CD123 (Cluster of Differentiation 123) is a 70-kD glycoprotein member of the hematopoietin receptor superfamily. This protein associates with a beta subunit common to the receptors for IL-5 and granulocyte-macrophage colony-stimulating factor (GM-CSF) to form a high-affinity receptor for IL-3. The interleukin-3 receptor α chain (CD123) has been identified as a potential immunotherapeutic target because it is overexpressed in AML compared with normal hematopoietic stem cells.