Learn More
ACROBiosystems ACROBiosystems Human IL-3 R alpha / CD123 Protein, Llama IgG2b Fc Tag, low endotoxin
Supplier:  ACROBiosystems ILAH5255100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IL-3 R alpha, Llama IgG2b Fc Tag, low endotoxin (ILA-H5255) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305  (Accession # P26951-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 60.9 kDa. The protein migrates as 45 kDa, 55 kDa and 80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6674