Learn More
ACROBiosystems Human IL-31 RA Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems ILAH52H7100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IL-31 RA, His Tag (ILA-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 519 (Accession # Q8NI17-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.0 kDa. The protein migrates as 90-115 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4808