Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-32 alpha ELISA Kit

Catalog No. EH274RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH274RBX5 5 x 96 Tests
EH274RBX10 10 x 96 Tests
EH274RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH274RBX5 Supplier Invitrogen™ Supplier No. EH274RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human IL-32 alpha quantitates human IL-32 alpha in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-32 alpha.

Interleukin 32 alpha (IL-32a) is a distinct isoform of the IL-32 cytokine, initially characterized as a transcript from activated NK cells. The gene encoding IL-32 is structured with eight exons, producing multiple alternatively spliced mRNA isoforms, including IL-32a. This isoform maintains similarity to other IL-32 variants, possessing sites for tyrosine sulfation, N-myristoylation, protein kinase C, casein kinase II phosphorylation, and RGD cell attachment, which are crucial for its functional interactions. IL-32a lacks a transmembrane region and is secreted, implicating a role in inflammation and immune processes by provoking pro-inflammatory cytokines while retaining certain anti-inflammatory features intracellularly. Particularly in the context of immune response, IL-32a is involved in defense mechanisms against intracellular pathogens and may have implications in cancerous processes, given its ability to influence cellular metabolism and proliferation. Its involvement in inflammation and immune regulation underlines its complex role in health and disease.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 4.1-1000 pg/mL
Assay Sensitivity 4.1 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias IL32; IL-32; IL-32alpha; IL-32beta; IL-32delta; IL-32gamma; interleukin 32; interleukin-32; interleukin-32 eta; interleukin-32 small; interleukin-32 theta; natural killer cell transcript 4; natural killer cells protein 4; NK4; TAIF; TAIFa; TAIFb; TAIFc; TAIFd; Tumor necrosis factor alpha-inducing factorIL32; IL-32; IL-32alpha; IL-32beta; IL-32delta; IL-32gamma; interleukin 32; interleukin-32; interleukin-32 eta; interleukin-32 small; interleukin-32 theta; natural killer cell transcript 4; natural killer cells protein 4; NK4; TAIF; TAIFa; TAIFb; TAIFc; TAIFd; Tumor necrosis factor alpha-inducing factor;
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IL-32 alpha
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.