Learn More
FUJIFILM BIOSCIENCES INC Human IL-32 alpha, Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 100151100UG
Encompass_Preferred
Interleukin 32 alpha (IL-32 A) is one of six known splice variants of the IL-32 gene. IL-32 A induces the macrophage production of inflammatory cytokines, such as interleukin 8 (IL-8), tumor necrosis factor-alpha (TNF-A), and macrophage inflammatory protein 2 (MIP-2). IL-32 A expression is increased after the activation of T cells, natural killer (NK) cells, and interferon gamma-treated epithelial cells.
Catalog No. 50-900-507