Learn More
FUJIFILM Biosciences Human IL-33, Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Catalog No. 50990684
Encompass_Preferred
Interleukin 33 (IL-33) is a member of the IL-1 cytokine family and is constitutively expressed in smooth muscle and airway epithelial cells. IL-33 signals through the interleukin 1 receptor-like 1 (IL-1R1) and interleukin-1 receptor accessory protein (IL1RAP) receptors to ativate NF-KB and MAPK signaling pathways. IL-33 functions to induce type 2 cytokine production in polarized Type 2 helper T (Th2) cells.
Catalog No. 50-990-684 Supplier FUJIFILM Biosciences Supplier No. 1001451MG
Add to Cart
Add to Cart
Request a Quote
Interleukin 33 (IL-33) is a member of the IL-1 cytokine family and is constitutively expressed in smooth muscle and airway epithelial cells. IL-33 signals through the interleukin 1 receptor-like 1 (IL-1R1) and interleukin-1 receptor accessory protein (IL1RAP) receptors to ativate NF-KB and MAPK signaling pathways. IL-33 functions to induce type 2 cytokine production in polarized Type 2 helper T (Th2) cells.