Learn More
R&D Systems™ Human IL-33 Quantikine ELISA Kit
Description
Specifications
Specifications
| Assay Range | 3.13 to 200 pg/mL |
| Assay Sensitivity | 1.51 pg/mL |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Tissue Lysates |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 90865 |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50uL),Cell Lysates (10uL),Tissue Lysates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | C9orf26, C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL33, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.