Learn More
Raybiotech Inc Human IL-34 ELISA Kit
Supplier:  Raybiotech Inc ELHIL341
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 7 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human ANGPTL3, beta IG-H3, CA9, Cathepsin B, CD23, CHI3L1, CTLA4, Dkk-4 , DPPIV, EDA-A2, Epo R, FGF-6, FGF-9, Gas1, IGFBP-5, IL-1F5, IL-1F6, IL-1F7, IL-1F8, IL-1F9, IL-1F10, IL-1R5, IL-17C, IL-18, IL-20, IL-5 R alpha, IL-10 R alpha, Layilin , Leptin R, Marapsin, Mer, MMP-7, P-Cadherin, Prostasin, PSMA, SIGIRR, TGF-beta RIII, Tissue Factor, TWEAK.
Catalog No. 50-194-9450